Learn More
Sino Biological Anti-Oct4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100616T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Oct4 / POU5F1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1682