Learn More
Sino Biological Anti-CAMKIV Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100746T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse CaM Kinase 4 / CaMKIV, and purified by antigen affinity chromatography.
Catalog No. 50-161-1971