Learn More
Sino Biological Recombinant Human PD-L1/B7-H1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10084H08H100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Thr 239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3965