Learn More
Sino Biological Anti-FBXO6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100914T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human FBXO6, and purified by antigen affinity chromatography.
Catalog No. 50-161-2370