Learn More
MINERVA BIOLABS INC 100CFU™ Sensitivity Standard, Spiroplasma citri
Supplier:  MINERVA BIOLABS INC 1039003
Encompass_Preferred
100CFU™ Sensitivity Standards are designed as complementary products to the 10CFU™ Sensitivity Standards (Cat. No. 102-XX03) to perform the validation of nucleic acid amplification technology (NAT)-based tests for mycoplasma detection.Content:3 vials containing 100 CFU of the particular mycoplasma as well as 2 vials negative control. The components are lyophilized.
Catalog No. NC3785858