Learn More
Cayman Chemical DPPH, 1898-66-4, 100 mg
SDP
Supplier:  Cayman Chemical 14805100
Encompass_Preferred
Molecular Formula C18H12N5O6, Purity ≥95%, Formula Weight 394.3, Synonyms: 2,2-diphenyl-1-(2,4,6-trinitrophenyl)-hydrazinyl, 2,2-Diphenyl-1-Picrylhydrazyl, DPPH radical; A cell-permeable, stable free radical that is commonly used to evaluate the ability of compounds to act as free radical scavengers or hydrogen donors and to measure the antioxidant activity of tissue extracts; followed by color change from purple (absorbance at 515-528 nm) to yellow.
Catalog No. NC0760517
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.