Learn More
XRF Scientific Americas Inc. PLATINUM EVAPORATION DISH FLAT BASE 100ML FROM XRF SCIENTIFIC AMERICAS
Supplier:  XRF Scientific Americas Inc. GED100
Encompass
67x28mm with a 100 ml capacity. Lids sold separately. These 99.95% pure platinum dishes have straight walls, and a flat base, & used for Fusions, Digestion, Ash testing, Loss on Ignition (LOI). due to their high temperature strenght, ductility, corrosion and oxidation, excellent resistance to chemical attacks by acids and fusion mixtures resistance; these platinum dishes are critical when high temperatures & resistance to chemical attacks are required.
Catalog No. NC1231716
 Restricted Item