Learn More
Biolegend Alexa Fluor 647 anti-human EGFR
Supplier:  Biolegend 352918
Encompass_Preferred
Alexa Fluor 647 anti-human EGFR [AY13]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1573462