Learn More
BIORBYT LLC NCadherinAntibody
Supplier:  BIORBYT LLC ORB1335506
Encompass
CategoryAntibodies.ClonalityMonoclonal.CloneOTI2G7.HostMouse.IsotypeIgG2a.MolecularWeight97.2kDa
Catalog No. NC2855328