Learn More
Abcam 100 x Citrate Buffer pH 6.0, 50ML
Supplier:  Abcam AB6423650ML
Encompass_Preferred
Citrate buffer pH 6.0 for heat-induced antigen retrieval (HIER) during IHC (ab64236).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1102437