Learn More
Sino Biological Recombinant Human VCAM-1/CD106 Protein (His Tag)
Supplier:  Sino Biological 10113H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human VCAM1 (NP_001069.1) (Met 1-Pro 697) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4040