Learn More
Sino Biological Recombinant Human JAML/AMICA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10120H08H100
Encompass_Preferred
A DNA sequence encoding the human JAML (NP_001091996.1) extracellular domain (Met 1-Leu 275) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4059