Learn More
Sino Biological Recombinant Human IL-1 alpha/IL1A Protein
Supplier:  Sino Biological 10128HNCH20
Encompass_Preferred
The processed form of human IL1α (NP_000566.3) (Ser 113-Ala 271) was expressed and purified.
Catalog No. 50-161-4092