Learn More
Sino Biological Recombinant Human tPA Protein (hFc Tag)
Supplier:  Sino Biological 10157H01H120
Encompass_Preferred
A DNA sequence encoding the α chain (Tyr 37-Arg 310) of mature human tPA (NP_000921.1) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4154