Learn More
Sino Biological Anti-Cystatin F/CST7 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10167MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CST7 (rh CST7; Catalog#10167-H08H; NP_003641.3; Met 1-His 145). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2337