Learn More
GREENHOUSE MEGASTORE 1020 TRAYS,HD,NO DRAIN,100/CS
SDP
Supplier:  GREENHOUSE MEGASTORE CNFLHDX2
Encompass
This heavy duty thermoformed tray is heavier than standard 1020 trays. Design allows community watering of pots, consistent bottom & side positioned drainage holes (or no holes) and easy, two-finger denesting. Both trays come with no center bar or side ribs. Select flats with or without holes.10.94" W x 21.44" L x 2.44" DThermoformed trays designed for consistent performance in automated filling and handling equipment.Don't settle for cheap and flimsy trays. Our 1020 trays are heavy weight and reusable.Used with our inserts to create compact, efficient growing systems for Bedding Plants5 oz eachStarting Thickness: 0.040" gauge (Heavy Duty)Made in the USA
Catalog No. NC2279954