Learn More
Sino Biological Recombinant Human E-Cadherin Protein (His Tag)
Supplier:  Sino Biological 10204H08H150
Encompass_Preferred
A DNA sequence encoding the human CDH1 (NP_004351.1) (Asp155-Ala709) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4326