Learn More
Sino Biological Recombinant Human LAYN Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10208H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 220) of human Layilin (NP_849156.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4334