Learn More
Sino Biological Recombinant Human Dectin-1/CLEC7A Protein, HPLC-verified
Supplier:  Sino Biological 10215HNCH100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of the human Dectin1 (NP_072092.2) (Thr66-Met201) was expressed.
Catalog No. 50-161-4360