Learn More
Sino Biological Recombinant Human MMP12 Protein (catalytic domain)
Supplier:  Sino Biological 10266HNAE20
Encompass_Preferred
A DNA sequence encoding human MMP12 (NP_002417.2) (Gly106-Asn268) was expressed with a N-terminal Met.
Catalog No. 50-161-4473