Learn More
Sino Biological Recombinant Human Neurotrophin 3/NT-3 Protein
Supplier:  Sino Biological 10286HNAE20
Encompass_Preferred
A DNA sequence encoding the human NT3 (P20783-1) (Tyr139-Thr257) was expressed and purified.
Catalog No. 50-161-4522