Learn More
Sino Biological Recombinant Human Fibronectin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10314H08H100
Encompass_Preferred
A DNA sequence encoding the Fragment 2 (Ser607-Pro1265) of human Fibronectin (NP_997647.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4580