Learn More
Sino Biological Anti-CDH12 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10317T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cadherin-12 / CDH12 (rh Cadherin-12 / CDH12; Catalog#10317-H08H; NP_001783.2; Met1-Ala605). Cadherin-12 / CDH12 specific IgG was purified by Human Cadherin-12 / CDH12 affinity chromatography.
Catalog No. 50-172-4415