Learn More
Sino Biological Anti-ICAM-3/CD50 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10333RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ICAM3 / CD50 (rh ICAM3 / CD50; Catalog#10333-H08H; CAA49369.1; Met 1-His 485). ICAM3 / CD50 specific IgG was purified by Human ICAM3 / CD50 affinity chromatography.
Catalog No. 50-172-4592