Learn More
Sino Biological Recombinant Human Iduronate 2 sulfatase/IDS Protein (His Tag)
Supplier:  Sino Biological 10337H08H50
Encompass_Preferred
A DNA sequence encoding human IDS precursor (NP_000193.1) (Met 1-Pro 550) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4646