Learn More
Sino Biological Recombinant Human SMAC Protein (His Tag)
Supplier:  Sino Biological 10339H08E100
Encompass_Preferred
A DNA sequence encoding the mitochondrial-located form of human Diablo (NP_063940.1) (Ala 56-Asp 239) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4651