Learn More
Sino Biological Recombinant Human Interferon alpha 10/IFNA10 Protein (hFc Tag)
Supplier:  Sino Biological 10349H01H100
Encompass_Preferred
A DNA sequence encoding the human IFNA10 (P01566) (Cys 24-Asp 189) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4689