Learn More
Sino Biological Recombinant Human CD32A/FCGR2A Protein (R167, hFc Tag)
Supplier:  Sino Biological 10374H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A (P12318-1, with Natural variant R167) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4763