Learn More
Sino Biological Recombinant Human CD32A/FCGR2A Protein (H167, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10374H27H1B20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Met210) of human FCGR2A (P12318-1, with Natural variant H167) was fused with a C-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-161-4778