Learn More
Sino Biological Anti-CD16/FCGR3/Fc gamma RIII Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10389MM22F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD16a/Fc gamma RIIIa (rh CD16a/Fc gamma RIIIa; Catalog#10389-H08H; P08637-1; Met1-Gln208,176F) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-5281