Learn More
Sino Biological Anti-PGD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10393T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PGD (rh PGD; Catalog#10393-H07E; P52209; Met1-Ala483). PGD specific IgG was purified by Human PGD affinity chromatography.
Catalog No. 50-172-5327