Learn More
Sino Biological Recombinant Human CD89/FCAR Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10414H08H100
Encompass_Preferred
A DNA sequence encoding the human FCAR (NP_001991.1) (Met1-Asn227) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4891