Learn More
Sino Biological Recombinant Human DcR1/TRAILR3 Protein (His Tag)
Supplier:  Sino Biological 10415H08H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF10C (NP_003832.2) (Met1-Pro235) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4893