Learn More
Sino Biological Recombinant Human ACYP2 Protein (GST Tag)
Supplier:  Sino Biological 10437H09E100
Encompass_Preferred
A DNA sequence encoding the human ACYP2 (P14621) (Ser 2-Tyr 99) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-4941