Learn More
Sino Biological Anti-CST6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10438RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cystatin-M / CST6 (rh Cystatin-M; Catalog#10438-H08H; NP_001314.1; Met 1-Met 149). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5814