Learn More
Sino Biological Recombinant Human CCL14 Protein (aa 28-93, His Tag)
Supplier:  Sino Biological 10476H07E150
Encompass_Preferred
A DNA sequence encoding the amino acids (Gly 28-Asn 93) of human CCL14 (Q16627-1) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5033