Learn More
Sino Biological Recombinant Human BCMA Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 10620H38H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF17 (NP_001183.2 ) (Met1-Ala54) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-161-5311