Learn More
Sino Biological Recombinant Human PLK1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10676H07B50
Encompass_Preferred
A DNA sequence encoding the human PLK1 (NP_005021.2) (Met 1-Ser 603) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5367