Learn More
Sino Biological Recombinant Human ULBP1/NKG2DL1 Protein (His & hFc Tag)
Supplier:  Sino Biological 10679H03H100
Encompass_Preferred
A DNA sequence encoding the mature form of human ULBP1 (NP_079494.1) (Met 1-Gly 216) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5373