Learn More
Sino Biological Recombinant Human B7-2/CD86 Protein (His Tag)
Supplier:  Sino Biological 10699H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-His 239) of human B7-2 (NP_008820.2) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5449