Learn More
Sino Biological Recombinant Human IFN-beta Protein
Supplier:  Sino Biological 10704HNAS20
Encompass_Preferred
A DNA sequence encoding the human IFNβ (P01574) (Met1-Asn187) was expressed.
Catalog No. 50-161-5483