Learn More
Sino Biological Recombinant Human Erythropoietin R/EPOR Protein (hFc Tag)
Supplier:  Sino Biological 10707H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Ala25-Pro250) of human erythropoietin receptor (NP_000112.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-5488