Learn More
Sino Biological Recombinant Human OXSR1 Protein (GST Tag)
Supplier:  Sino Biological 10727H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human OXSR1 (NP_005100.1) (Met 1-Ser 527) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5507