Learn More
Sino Biological Recombinant Human P38 Delta/MAPK13 Protein (GST Tag), Activated in vitro
Supplier:  Sino Biological 10747H09BA50
Encompass_Preferred
A DNA sequence encoding the full length of human MAPK13 (O15264) (Met1-Leu365) was fused with the GST tag at the N-terminus. Activated in vitro by MAP2K6 (10422-H20B1).
Catalog No. 50-161-5538