Learn More
Sino Biological Recombinant Human CXCL5 Protein
Supplier:  Sino Biological 10889HNAE20
Encompass_Preferred
A DNA sequence encoding the human CXCL5 (P42830) (Arg45-Asn114) was expressed, with a N-terminal Met.
Catalog No. 50-161-5791