Learn More
Oxyrase Oxyblue Indicator (10 Bulbs Per Pack) 10/Pk
SDP
Supplier:  Oxyrase AOX0010
Encompass
Anaerobic indicator which changes color repeatedly based on environment. Indicator turns white when environment becomes anaerobic. Indicator turns back to blue when exposed to ambient oxygen. Convenient applicator. Affordable.
Catalog No. 50-200-4799