Learn More
Atlantic Scale Co 10 kg Calibration Weight, ASTM Class 1, Includes Certificate, 10 kg
SDP
Supplier:  Atlantic Scale Co TRO80781003
Encompass
10 KG CALIBRATION WEIGHT W/CERT is a precision-crafted instrument designed for scientific and laboratory applications requiring high accuracy. Made from 303 stainless steel, it features a polished finish and a recessed grip for secure handling. This cylindrical weight ensures compliance with ASTM Class 1 standards and is supplied with a certification of accuracy by ATLANTIC SCALE COMPANY INC, setting it apart from others typically offered without certification.
Catalog No. NC3597599