Learn More
Sentry Air Systems Model 300 Activated Carbon Filter - 10 lb
SDP
Supplier:  Sentry Air Systems SS310CF
Encompass
Model 300 Activated Carbon Filter features 10 lbs of heavy-duty activated carbon from crushed virgin coconuts. Carbon filters are ideal for the removal of gases, fumes, and vapors. Efficiency depends on the application. Fits Model 300 systems.
Catalog No. NC9989577