Learn More
Selleck Chemical LLC StemRegenin 1 (SR1) 10mg 1227633-49-9
SDP
Supplier:  Selleck Chemical LLC S285810MG
Encompass_Preferred
StemRegenin 1 (SR1) is an aryl hydrocarbon receptor (AhR) inhibitor with IC50 of 127 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1216744