Learn More
S2 Media 10mL 2% Brilliant Green Bile Broth (BGBB), 12mL Polypropylene Flat Bottom Tube with Inverted Durham Tube, 50/box
Supplier:  S2 Media 5113
Encompass_Preferred
Brilliant Green Bile Broth is a selective broth used for the qualitative detection of coliform bacteria. Coliforms include aerobic and facultative anaerobic, Gram-negative, non-spore forming bacilli that ferment lactose and form acid and gas at 35°C within 48 hours. Members of Enterobacteriaceae comprise the majority of this group, but organisms such as Aeromonas spp. may also be included. Brilliant Green Bile Broth is used for confirmation testing of positive presumptive test results. • Product meets APHA, AOAC, BAM, EPA, ISO, SM-Dairy, and SMWW reference methods and performance criteria.• BBGB should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Brilliant Green Bile Broth 2% and Brilliant Green Lactose Bile Broth.
Catalog No. 50-299-5954